1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper
1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper
1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper
1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper
Thermo Scientific Chemicals

1-Bromo-2-iodotetrafluoroethane, 97%, stab. with copper

CAS: 421-70-5 | C2BrF4I | 306.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21771.06
also known as B21771-06
Price (USD)
75.65
Online Exclusive
84.40
Save 8.75 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
84.40
Save 8.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS421-70-5
IUPAC Name1-bromo-1,1,2,2-tetrafluoro-2-iodoethane
Molecular FormulaC2BrF4I
InChI KeyZYNPYKGTNSXKPI-UHFFFAOYSA-N
SMILESFC(F)(Br)C(F)(F)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Assay (GC)>96.0%
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only