2-Chlorobutyramide, 98%, Thermo Scientific Chemicals
2-Chlorobutyramide, 98%, Thermo Scientific Chemicals
2-Chlorobutyramide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chlorobutyramide, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B21783.06
also known as B21783-06
5 g
Catalog number B21783.06
also known as B21783-06
Price (USD)
80.65
Online Exclusive
89.70
Save 9.05 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
80.65
Online Exclusive
89.70
Save 9.05 (10%)
Each
Chemical Identifiers
CAS7462-73-9
IUPAC Name2-chlorobutanamide
Molecular FormulaC4H8ClNO
InChI KeyVJIOSCMTZVXQQU-UHFFFAOYNA-N
SMILESCCC(Cl)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)73.0-79.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only