Polyethylene glycol 600
Polyethylene glycol 600
Polyethylene glycol 600
Thermo Scientific Chemicals

Polyethylene glycol 600

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21798.0B
also known as B21798-0B
Price (USD)
86.65
Online Exclusive
96.40
Save 9.75 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.40
Save 9.75 (10%)
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
FormLiquid as melt or white low-melting solid
pH4.5-7.5 (5%, demin water)
Hydroxyl Value178-197 mg KOH/g
Appearance (Color)Colorless
CommentAverage mol weight (based on OH value) 570-630
Polyethylene glycol is used as a humectant in inks and adhesives. It acts as a solvent in soldering flux and as a dye carrier in inks. It is also useful as a modifier and binder in latex paint. It is used as an additive in synthetic lubricants, cutting oil and hydraulic fluids. It plays an important role for the precipitation of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only