Vinyl benzoate, 95%
Vinyl benzoate, 95%
Vinyl benzoate, 95%
Thermo Scientific Chemicals

Vinyl benzoate, 95%

CAS: 769-78-8 | C9H8O2 | 148.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B21805.22
also known as B21805-22
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.10
Save 9.45 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.10
Save 9.45 (14%)
Each
Chemical Identifiers
CAS769-78-8
IUPAC Nameethenyl benzoate
Molecular FormulaC9H8O2
InChI KeyKOZCZZVUFDCZGG-UHFFFAOYSA-N
SMILESC=COC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test10% in diethyl ether: Clear
FormLiquid
Refractive Index1.5250-1.5310 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥94.0%
View more
It is used as chemical reagent and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as chemical reagent and pharmaceutical intermediates.

Solubility
Soluble in methanol

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Mikiharu Kamachi.; Jiro Satoh.; Der Jang Liaw.; Shun-ichi Nazakura. Solvent Effects on Radical Polymerization of Vinyl Benzoate and Phenyl Methacrylate.Macromolecules. 1977, 10 (2),501-502.
  2. Yoshinori Kamiya;Keishin Mizoguchi.; Yasutoshi Naito.; Takuji Hirose. Gas sorption in poly(vinyl benzoate). J. Polym. Sci., Part B: Polym. Phys. 1986, 24 (3),535-547.