4-Cyano-4'-n-pentylbiphenyl, 99%
4-Cyano-4'-n-pentylbiphenyl, 99%
4-Cyano-4'-n-pentylbiphenyl, 99%
Thermo Scientific Chemicals

4-Cyano-4'-n-pentylbiphenyl, 99%

CAS: 40817-08-1 | C18H19N | 249.357 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B21856.06
also known as B21856-06
Price (USD)
248.65
Online exclusive
276.00
Save 27.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
248.65
Online exclusive
276.00
Save 27.35
Each
Chemical Identifiers
CAS40817-08-1
IUPAC Name4'-pentyl-[1,1'-biphenyl]-4-carbonitrile
Molecular FormulaC18H19N
InChI KeyHHPCNRKYVYWYAU-UHFFFAOYSA-N
SMILESCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or clear liquid as melt
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Refractive Index1.5315-1.5345 @ 20?C
Appearance (Color)Colorless to white
4-Cyano-4'-n-pentylbiphenyl is used in the preparation of 4'-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-4′-n-pentylbiphenyl is used in the preparation of 4′-pentylbiphenyl-4-carboxylic acid by acidic hydrolysis using sulfuric acid as a reagent.

Solubility
Miscible with Acetone, chloroform, toluene, acetonitrile, methylene chloride. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Matsushita, S.; Yan, B.; Yamamoto, S.; Jeong, Y. S.; Akagi, K. Helical Carbon and Graphite Films Prepared from Helical Poly(3,4-ethylenedioxythiophene) Films Synthesized by Electrochemical Polymerization in Chiral Nematic Liquid Crystals. Angew. Chem. Int. Ed. 2014, 53 (6), 1659-1663.
  2. Kasch, N.; Dierking, I.; Turner, M.; Romero-Hasler, P.; Soto-Bustamante, E. A. Liquid crystalline textures and polymer morphologies resulting from electropolymerisation in liquid crystal phases. J. Mater. Chem. C 2015, 3 (31), 8018-8023.