2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%
2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%
2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%
Thermo Scientific Chemicals

2-(4-Methyl-5-thiazolyl)ethyl acetate, 98+%

CAS: 656-53-1 | C8H11NO2S | 185.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21867.14
also known as B21867-14
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS656-53-1
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5080-1.5120 @ 20?C
Appearance (Color)Clear pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only