3-Phenoxytoluene, 97%
3-Phenoxytoluene, 97%
3-Phenoxytoluene, 97%
Thermo Scientific Chemicals

3-Phenoxytoluene, 97%

CAS: 3586-14-9 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21899.14
also known as B21899-14
Price (USD)
38.65
Online exclusive
42.60
Save 3.95
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
38.65
Online exclusive
42.60
Save 3.95
Each
Chemical Identifiers
CAS3586-14-9
IUPAC Name1-methyl-3-phenoxybenzene
Molecular FormulaC13H12O
InChI KeyUDONPJKEOAWFGI-UHFFFAOYSA-N
SMILESCC1=CC=CC(OC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5710-1.5760 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only