Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate
Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate
Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate
Thermo Scientific Chemicals

Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate

CAS: 1191-50-0 | C14H29NaO4S | 316.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21941.06
also known as B21941-06
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS1191-50-0
IUPAC Namesodium tetradecyl sulfate
Molecular FormulaC14H29NaO4S
InChI KeyUPUIQOIQVMNQAP-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Supplier's CofA≥94.0%
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Fernandez-Ramos, C.; Ballesteros, O.; Zafra-Gomez, A.; Blanc, R.; Navalon, A.; Vílchez, J. L. Determination of alcohol sulfates and alcohol ethoxysulfates in wastewater samples by liquid chromatography tandem mass spectrometry. Microchem. J. 2013, 106, 180-185.
  2. Fernandez-Ramos, C.; Ballesteros, O.; Blanc, R.; Zafra-Gomez, A.; Jimenez-Díaz, I.; Navalón, A.; Vílchez, J. L. Determination of alcohol sulfates in wastewater treatment plant influents and effluents by gas chromatography-mass spectrometry. Talanta 2012, 98, 166-171.