Phenol Red sodium salt
Phenol Red sodium salt
Phenol Red sodium salt
Thermo Scientific Chemicals

Phenol Red sodium salt

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B21948.14
also known as B21948-14
Price (USD)
83.80
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
83.80
Each
Chemical Identifiers
CAS34487-61-1
IUPAC Namesodium 4-[3-(4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular FormulaC19H13NaO5S
InChI KeyHKHYOKBQJILTEI-UHFFFAOYNA-M
SMILES[Na+].OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
CommentMaterial sourced in UK and US
Color ChangeYellow @ pH ≤6.8, Red @ pH ≥8.2
Solution Test1 mg/L in water (pH 7): Clear orange solution (UK-sourced material)
Appearance (Color)Red to dark red
Phenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.

Solubility
Soluble in water yielding a dark red solution (25 mg/ml), alcohol (4 mg/ml), methanol, ethanol (200 mg/ml), and aqueous alkali hydroxides or carbonates with formation of a red colored solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
F. Alan McDonald,; Grover C. Wetsel Jr. Generalized theory of the photoacoustic effect. J. Appl. Phys. 1978, 49 2313.