1-Phenylnaphthalene, 97%
1-Phenylnaphthalene, 97%
1-Phenylnaphthalene, 97%
Thermo Scientific Chemicals

1-Phenylnaphthalene, 97%

CAS: 605-02-7 | C16H12 | 204.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B21953.22
also known as B21953-22
Price (USD)
736.65
Special offer
Online exclusive
Ends: 30-Jun-2026
867.00
Save 130.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
736.65
Special offer
Online exclusive
Ends: 30-Jun-2026
867.00
Save 130.35 (15%)
Each
Chemical Identifiers
CAS605-02-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.6635-1.6685 @ 20°C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only