2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water
2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water
2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water
Thermo Scientific Chemicals

2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water

CAS: 886-38-4 | C15H10O | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B21954.03
also known as B21954-03
Price (USD)
57.65
Online Exclusive
63.50
Save 5.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.50
Save 5.85 (9%)
Each
Chemical Identifiers
CAS886-38-4
IUPAC Namediphenylcycloprop-2-en-1-one
Molecular FormulaC15H10O
InChI KeyHCIBTBXNLVOFER-UHFFFAOYSA-N
SMILESO=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale orange or orange-pink
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤10%
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only