Polyethylene glycol 10,000
Polyethylene glycol 10,000
Polyethylene glycol 10,000
Thermo Scientific Chemicals

Polyethylene glycol 10,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B21955.30
Price (USD)
47.30
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
47.30
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Solution TestClear, APHA ≤30
Viscosity550 - 750 mPas (50%, water @ 20?C)
Appearance (Color)White
FormPowder or crystals or flakes
CommentAveraged mol weight 9000 - 11250 (based on OH value)
View more
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only