2-Octynoic acid, 98%
2-Octynoic acid, 98%
2-Octynoic acid, 98%
Thermo Scientific Chemicals

2-Octynoic acid, 98%

CAS: 5663-96-7 | C8H12O2 | 140.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number B21957.06
also known as B21957-06
Price (USD)
99.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
99.10
Each
Chemical Identifiers
CAS5663-96-7
IUPAC Nameoct-2-ynoic acid
Molecular FormulaC8H12O2
InChI KeyBQDKCWCMDBMLEH-UHFFFAOYSA-N
SMILESCCCCCC#CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.4590-1.4630 @ 20°C
FormLiquid
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only