2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%
2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%
2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%
Thermo Scientific Chemicals

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%

CAS: 35011-47-3 | C4H9N5O5S | 239.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B21989.22
also known as B21989-22
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS35011-47-3
IUPAC Name2,5,6-triamino-1,4-dihydropyrimidin-4-one; sulfuric acid
Molecular FormulaC4H9N5O5S
InChI KeyRSKNEEODWFLVFF-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC(=O)C(N)=C(N)N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale yellow to yellow or pale brown
Acid-base back titration≥92.0 to ≤108.0%
Identification (FTIR)Conforms
Assay (HPLC)≥92.0%
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S Kaufman. The activity of 2,4,5-triamino-6-hydroxypyrimidine in the phenylalanine hydroxylase system. The Journal of Biological Chemistry. 1979, 254, 5150-5154.
  2. H. S. Forrest and James Walker. The effect of hydrazine on the condensation of certain α-ketols and related substances with 2 : 4 : 5-triamino-6-hydroxypyrimidine. J. Chem. Soc. 1949, 2077-2082.