2-Methyl-5-(methylthio)furan, 99%
2-Methyl-5-(methylthio)furan, 99%
2-Methyl-5-(methylthio)furan, 99%
Thermo Scientific Chemicals

2-Methyl-5-(methylthio)furan, 99%

CAS: 13678-59-6 | C6H8OS | 128.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number B21998.04
also known as B21998-04
Price (USD)
47.65
Online Exclusive
53.10
Save 5.45 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
53.10
Save 5.45 (10%)
Each
Chemical Identifiers
CAS13678-59-6
IUPAC Name2-methyl-5-(methylsulfanyl)furan
Molecular FormulaC6H8OS
InChI KeyRESBOJMQOGJOMW-UHFFFAOYSA-N
SMILESCSC1=CC=C(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive Index1.5165-1.5195 @ 20?C
Assay (GC)≥98.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only