1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%
Thermo Scientific Chemicals

1-(4-Hydroxyphenyl)-5-mercaptotetrazole, 96%

CAS: 52431-78-4 | C7H6N4OS | 194.212 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22003.06
also known as B22003-06
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.30
Save 4.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.30
Save 4.65 (15%)
Each
Chemical Identifiers
CAS52431-78-4
IUPAC Name1-(4-hydroxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Molecular FormulaC7H6N4OS
InChI KeyMOXZSKYLLSPATM-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)N1NN=NC1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to purple
FormCrystals or powder or crystalline powder or pieces
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0% (dry wt. basis)
Assay (HPLC)≥95.0%
Loss on Drying≤5.0% (105°C/constant weight)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only