Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Mesitylenesulfonic acid sodium salt hemihydrate, 98%
Thermo Scientific Chemicals

Mesitylenesulfonic acid sodium salt hemihydrate, 98%

CAS: 6148-75-0 | C9H11NaO3S | 222.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22029.14
also known as B22029-14
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS6148-75-0
SpecificationsSpecification SheetSpecification Sheet
Assay (ex Sulphated Ash)≥97.5 to ≤102.5% (as hemihydrate)
Appearance (Color)White to Cream or Pale yellow
Water Content (Karl Fischer Titration)3.0-5.0% (0.38-0.65 waters of hydration)
Sulfated Ash29.94-31.48% (as hemihydrate; Theory 30.71%)
FormCrystals or powder or crystalline powder or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only