Thermo Scientific Chemicals

Mesitylenesulfonic acid sodium salt hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: B22029.22
100 g, Each
Thermo Scientific Chemicals

Mesitylenesulfonic acid sodium salt hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: B22029.22
100 g, Each
Quantity
Catalog number: B22029.22
also known as B22029-22
Price (USD)
Price: 176.00
Online price: 149.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6148-75-0
Water Content (Karl Fischer Titration)
3.0-5.0% (0.38-0.65 waters of hydration)
Sulfated Ash
29.94-31.48% (as hemihydrate; Theory 30.71%)
Appearance (Color)
White to Cream or Pale yellow
Assay (ex Sulphated Ash)
≥97.5 to ≤102.5% (as hemihydrate)
Form
Crystals or powder or crystalline powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text