Trimethylsilyl acetate, 97%
Trimethylsilyl acetate, 97%
Trimethylsilyl acetate, 97%
Thermo Scientific Chemicals

Trimethylsilyl acetate, 97%

CAS: 2754-27-0 | C5H12O2Si | 132.234 g/mol
Have Questions?
Catalog NumberQuantity
B22037.22
also known as B22037-22
100 g
Catalog number B22037.22
also known as B22037-22
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
83.65
Online Exclusive
92.30
Save 8.65 (9%)
Each
Chemical Identifiers
CAS2754-27-0
IUPAC Nametrimethylsilyl acetate
Molecular FormulaC5H12O2Si
InChI KeyQHUNJMXHQHHWQP-UHFFFAOYSA-N
SMILESCC(=O)O[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3855-1.3905 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only