1,2,3-Trifluorobenzene, 98+%
1,2,3-Trifluorobenzene, 98+%
1,2,3-Trifluorobenzene, 98+%
Thermo Scientific Chemicals

1,2,3-Trifluorobenzene, 98+%

CAS: 1489-53-8 | C6H3F3 | 132.085 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22055.03
also known as B22055-03
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS1489-53-8
IUPAC Name1,2,3-trifluorobenzene
Molecular FormulaC6H3F3
InChI KeyAJKNNUJQFALRIK-UHFFFAOYSA-N
SMILESFC1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.4220-1.4260 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only