Pyrazine-2,3-dicarboxylic acid, 98%
Pyrazine-2,3-dicarboxylic acid, 98%
Pyrazine-2,3-dicarboxylic acid, 98%
Thermo Scientific Chemicals

Pyrazine-2,3-dicarboxylic acid, 98%

CAS: 89-01-0 | C6H4N2O4 | 168.108 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22108.06
also known as B22108-06
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS89-01-0
IUPAC Namepyrazine-2,3-dicarboxylic acid
Molecular FormulaC6H4N2O4
InChI KeyZUCRGHABDDWQPY-UHFFFAOYSA-N
SMILESOC(=O)C1=NC=CN=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only