Polyethylene glycol 1,000
Polyethylene glycol 1,000
Polyethylene glycol 1,000
Thermo Scientific Chemicals

Polyethylene glycol 1,000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number B22134.0B
also known as B22134-0B
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
FormWaxy solid
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Hydroxyl Value107-118 mg KOH/g
Appearance (Color)White
View more
Polyethylene glycol is used as an excipent in pharmaceutical products. It is used in the precipitation of proteins as well as in the separation and purification of biomolecules and in the induction of cell hybridization. It acts as a fusing agent to enhance the effect of macrophages on hybridoma; as a vascular agent in preclinical work; as an anti-foaming agent in food and as the gate insulator in an electric double-layer transistor to enhance superconductivity in an insulator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only