S-Methyl thiobutyrate, 98%
S-Methyl thiobutyrate, 98%
S-Methyl thiobutyrate, 98%
Thermo Scientific Chemicals

S-Methyl thiobutyrate, 98%

CAS: 2432-51-1 | C5H10OS | 118.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22144.06
also known as B22144-06
Price (USD)
36.65
Online exclusive
40.20
Save 3.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
36.65
Online exclusive
40.20
Save 3.55
Each
Chemical Identifiers
CAS2432-51-1
IUPAC Name1-(methylsulfanyl)butan-1-one
Molecular FormulaC5H10OS
InChI KeyGRLJIIJNZJVMGP-UHFFFAOYSA-N
SMILESCCCC(=O)SC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4605-1.4645 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only