4-Cyanophenylhydrazine hydrochloride, 97%
4-Cyanophenylhydrazine hydrochloride, 97%
4-Cyanophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

4-Cyanophenylhydrazine hydrochloride, 97%

CAS: 2863-98-1 | C7H7N3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22157.22
also known as B22157-22
Price (USD)
700.65
Online Exclusive
778.00
Save 77.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
700.65
Online Exclusive
778.00
Save 77.35 (10%)
Each
Chemical Identifiers
CAS2863-98-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink or pale brown
FormPowder
Assay (Titration ex Chloride)>96.0 to <104.0%
Assay (Freebase GC)>96.0%
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Latifeh Navidpour; Mohsen Amini; Hamed Shafaroodi; Khosrou Abdi; Mohammad H. Ghahremanic, Ahmad Reza Dehpourd, Abbas Shafiee. Design and synthesis of new water-soluble tetrazolide derivatives of celecoxib and rofecoxib as selective cyclooxygenase-2 (COX-2) inhibitors. Bioorganic & Medicinal Chemistry Letters. 2006, 16 (17), 4483-4487.
  2. Erzsébet Mernyák; László Márk; Éva Frank; Gyula Schneider; János Wölfling. Electrophile-induced generation of cyclic azomethine imines from steroidal δ-alkenyl hydrazones. Steroids. 2009, 74 (4-5), 474-482.