N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals
N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals
N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals
N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-tyrosine methyl ester, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
B22164.22
also known as B22164-22
100g
B22164.06
also known as B22164-06
5g
B22164.14
also known as B22164-14
25g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B22164.22
also known as B22164-22
Price (USD)
-
Quantity:
100g
Request bulk or custom format
Chemical Identifiers
CAS4326-36-7
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC15H21NO5
InChI KeyNQIFXJSLCUJHBB-STGVRZAANA-N
SMILESCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)103.0-109.0°C
Optical Rotation49.9 ± 1.0° (C=1 in chloroform)
FormPowder
Appearance (Color)White
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only