1-Methylpiperidine-2-methanol, 97%
1-Methylpiperidine-2-methanol, 97%
1-Methylpiperidine-2-methanol, 97%
Thermo Scientific Chemicals

1-Methylpiperidine-2-methanol, 97%

CAS: 20845-34-5 | C7H15NO | 129.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22187.14
also known as B22187-14
Price (USD)
71.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
71.60
Each
Chemical Identifiers
CAS20845-34-5
IUPAC Name(1-methylpiperidin-2-yl)methanol
Molecular FormulaC7H15NO
InChI KeyHXXJMMLIEYAFOZ-UHFFFAOYNA-N
SMILESCN1CCCCC1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Refractive Index1.4805-1.4855 @ 20°C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only