6-Methoxy-2-methylquinoline, 97%
6-Methoxy-2-methylquinoline, 97%
6-Methoxy-2-methylquinoline, 97%
6-Methoxy-2-methylquinoline, 97%
Thermo Scientific Chemicals

6-Methoxy-2-methylquinoline, 97%

CAS: 1078-28-0 | C11H11NO | 173.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22199.03
also known as B22199-03
Price (USD)
54.65
Online Exclusive
60.30
Save 5.65 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.30
Save 5.65 (9%)
Each
Add to cart
Chemical Identifiers
CAS1078-28-0
IUPAC Name6-methoxy-2-methylquinoline
Molecular FormulaC11H11NO
InChI KeyNAGJQQFMJKMXJQ-UHFFFAOYSA-N
SMILESCOC1=CC=C2N=C(C)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)59.0-69.0?C
Appearance (Color)Pale cream to cream to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only