Sodium 1-propanesulfonate, 99%
Sodium 1-propanesulfonate, 99%
Sodium 1-propanesulfonate, 99%
Thermo Scientific Chemicals

Sodium 1-propanesulfonate, 99%

CAS: 14533-63-2 | C3H7NaO3S | 146.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22210.03
also known as B22210-03
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
Chemical Identifiers
CAS14533-63-2
IUPAC Namesodium propane-1-sulfonate
Molecular FormulaC3H7NaO3S
InChI KeyNPAWNPCNZAPTKA-UHFFFAOYSA-M
SMILES[Na+].CCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)≥98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)<1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only