n-Butyl chloroformate, 98%
n-Butyl chloroformate, 98%
n-Butyl chloroformate, 98%
Thermo Scientific Chemicals

n-Butyl chloroformate, 98%

CAS: 592-34-7 | C5H9ClO2 | 136.575 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Catalog number B22215.22
also known as B22215-22
Price (USD)
38.40
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
38.40
Each
Chemical Identifiers
CAS592-34-7
IUPAC Namebutyl carbonochloridate
Molecular FormulaC5H9ClO2
InChI KeyNRDQFWXVTPZZAZ-UHFFFAOYSA-N
SMILESCCCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4110-1.4150 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only