4'-n-Octyloxybiphenyl-4-carboxylic acid, 99%
4'-n-Octyloxybiphenyl-4-carboxylic acid, 99%
4'-n-Octyloxybiphenyl-4-carboxylic acid, 99%
Thermo Scientific Chemicals

4'-n-Octyloxybiphenyl-4-carboxylic acid, 99%

CAS: 59748-18-4 | C21H26O3 | 326.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22216.03
also known as B22216-03
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.60
Save 5.95
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
43.60
Save 5.95
Each
Chemical Identifiers
CAS59748-18-4
IUPAC Name4'-(octyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Molecular FormulaC21H26O3
InChI KeyYNBBQLUKHHSKPW-UHFFFAOYSA-N
SMILESCCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only