1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%
1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%
1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%
Thermo Scientific Chemicals

1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%

CAS: 88462-65-1 | C11H9F3O2 | 230.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22218.06
also known as B22218-06
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
159.65
Online Exclusive
177.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS88462-65-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)29.0-35.0?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only