Acid Fuchsin sodium salt
Acid Fuchsin sodium salt
Acid Fuchsin sodium salt
Thermo Scientific Chemicals

Acid Fuchsin sodium salt

CAS: 3244-88-0 | C20H19N3Na2O9S3 | 587.544 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B22222.14
also known as B22222-14
Price (USD)
70.65
Online Exclusive
78.10
Save 7.45 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS3244-88-0
IUPAC Namedisodium 2-amino-5-[(4-amino-3-sulfophenyl)[(1Z)-4-imino-3-methyl-5-sulfonatocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular FormulaC20H17N3Na2O9S3
InChI KeyWZRZTHMJPHPAMU-UHFFFAOYSA-L
SMILES[Na+].[Na+].CC1=C\C(C=C(C1=N)S([O-])(=O)=O)=C(\C1=CC=C(N)C(=C1)S(O)(=O)=O)C1=CC=C(N)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark brown
FormCrystals or powder or crystalline powder
lambda-max (UV-Vis)540 - 550nm (water)
Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acid Fuchsin sodium salt is used as a pH indicator. It is one of the dyes used in Masson’s Trichome Stain where it is used to distinguish muscle from collagen.

Solubility
Soluble in water (1 mg/ml).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. W. L. Holman. The Use of Decolorized Acid Fuchsin as an Acid Indicator in Carbohydrate Fermentation Tests with Some Remarks on Acid Production by Bacteria. The Journal of Infectious Diseases. 1914, 15 (1), 227-233.
  2. Lene Feldskov Nielsena; Dennis Moe; Svend Kirkeby & Charly Garbarsch. BSirius Red and Acid Fuchsin Staining Mechanisms. Biotechnic & Histochemistry 1998, 73 (2), 71-77.