Sodium nicotinate, 98%
Sodium nicotinate, 98%
Sodium nicotinate, 98%
Thermo Scientific Chemicals

Sodium nicotinate, 98%

CAS: 54-86-4 | C6H4NNaO2 | 145.093 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22224.22
also known as B22224-22
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Chemical Identifiers
CAS54-86-4
IUPAC Namesodium pyridine-3-carboxylate
Molecular FormulaC6H4NNaO2
InChI KeyKFLRWGSAMLBHBV-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point>300?C
FormPowder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only