3-Methyl-2-butenal, 97%
3-Methyl-2-butenal, 97%
3-Methyl-2-butenal, 97%
3-Methyl-2-butenal, 97%
Thermo Scientific Chemicals

3-Methyl-2-butenal, 97%

CAS: 107-86-8 | C5H8O | 84.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22226.06
also known as B22226-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.00
Save 10.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
69.00
Save 10.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS107-86-8
IUPAC Name3-methylbut-2-enal
Molecular FormulaC5H8O
InChI KeySEPQTYODOKLVSB-UHFFFAOYSA-N
SMILESCC(C)=CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4590-1.4650 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
View more
3-Methyl-2-butenal is used in small quantities as food flavor in alcoholic beverages, chewing gums, confection frosting, frozen dairy, fruit ice, gelatin pudding, hard and soft candy and jam jelly. Used in daily flavor, and cosmetic flavor in perfumes. A starting material for the synthesis of vitamin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-butenal is used in small quantities as food flavor in alcoholic beverages, chewing gums, confection frosting, frozen dairy, fruit ice, gelatin pudding, hard and soft candy and jam jelly. Used in daily flavor, and cosmetic flavor in perfumes. A starting material for the synthesis of vitamin A.

Solubility
Soluble in water (110g/L).

Notes
Air sensitive. Store away from strong oxidizing agents, air, heat.
RUO – Research Use Only

General References:

  1. Brian G. Brownlee; Ross H. Hall; C. Dennis Whitty. 3-Methyl-2-butenal: An Enzymatic Degradation Product of the Cytokinin, N6-(Δ2-Isopentenyl)adenine. Canadian Journal of Biochemistry. 1975, 53 (1), 37-41.
  2. Akshay Waghray; Jian Wang, Rachid Oukaci; Donna G. Blackmond. Influence of alkali promoters in the selective hydrogenation of 3-methyl-2-butenal over ruthenium/silica catalysts. J. Phys. Chem. 1992, 96 (14), 5954-5959.