2-Octen-4-one, 97%
2-Octen-4-one, 97%
2-Octen-4-one, 97%
Thermo Scientific Chemicals

2-Octen-4-one, 97%

CAS: 4643-27-0 | C8H14O | 126.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22250.03
also known as B22250-03
Price (USD)
78.65
Online exclusive
87.50
Save 8.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
78.65
Online exclusive
87.50
Save 8.85
Each
Chemical Identifiers
CAS4643-27-0
IUPAC Name(2E)-oct-2-en-4-one
Molecular FormulaC8H14O
InChI KeyFMDLEUPBHMCPQV-GQCTYLIASA-N
SMILESCCCCC(=O)C=CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4425-1.4475 @ 20?C
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the rhodium-catalyzed asymmetric ring opening of oxabicyclic alkenes with organoboronic acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Ate Duursma,; Rob Hoen,; Julia Schuppan,; Ron Hulst,; Adriaan J. Minnaard,; Ben L. Feringa. First Examples of Improved Catalytic Asymmetric C-C Bond Formation Using the Monodentate Ligand Combination Approach. Journal name. 2003 volume (17), 3111-3113.