Ranitidine hydrochloride, 99%
Ranitidine hydrochloride, 99%
Ranitidine hydrochloride, 99%
Thermo Scientific Chemicals

Ranitidine hydrochloride, 99%

CAS: 66357-59-3 | C13H23ClN4O3S | 350.862 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B22260.06
also known as B22260-06
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
Chemical Identifiers
CAS66357-59-3
IUPAC Namehydrogen dimethyl[(5-{[(2-{[(1E)-1-(methylamino)-2-nitroethenyl]amino}ethyl)sulfanyl]methyl}furan-2-yl)methyl]amine chloride
Molecular FormulaC13H23ClN4O3S
InChI KeyGGWBHVILAJZWKJ-KJEVSKRMSA-N
SMILES[H+].[Cl-].CNC(NCCSCC1=CC=C(CN(C)C)O1)=C/[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
FormPowder
An H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An H-2 histamine receptor antagonistRanitidine hydrochloride is mainly used as an H2-receptor antagonist and helps in the treatment of gastrointestinal lesions because of excessive gastric acid secretion. It also serves as an antiulcer drug.

Solubility
Soluble in water, 2-hydroxypropyl-beta-cyclodextrin, acetic acid, methanol, ethanol and dimethyl sulfoxide. Insoluble in chloroform.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mondal, S.; Sinha, K.; Aikat, K.; Halder, G. Adsorption thermodynamics and kinetics of ranitidine hydrochloride onto superheated steam activated carbon derived from mung bean husk. J. Environ. Chem. Eng. 2015, 3 (1), 187-195.
  2. Trygg, J.; Yildir, E.; Kolakovic, R.; Sandler, N.; Fardim, P. Solid-State Properties and Controlled Release of Ranitidine Hydrochloride from Tailored Oxidised Cellulose Beads. Macromol. Mater. Eng. 2015, 300 (2), 210-217.