p-Mentha-8-thiol-3-one, cis + trans, 97%
p-Mentha-8-thiol-3-one, cis + trans, 97%
p-Mentha-8-thiol-3-one, cis + trans, 97%
p-Mentha-8-thiol-3-one, cis + trans, 97%
Thermo Scientific Chemicals

p-Mentha-8-thiol-3-one, cis + trans, 97%

CAS: 38462-22-5 | C10H18OS | 186.313 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22287.14
also known as B22287-14
Price (USD)
194.65
Special offer
Online exclusive
Ends: 31-Dec-2025
229.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 31-Dec-2025
229.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS38462-22-5
IUPAC Name5-methyl-2-(2-sulfanylpropan-2-yl)cyclohexan-1-one
Molecular FormulaC10H18OS
InChI KeyRVOKNSFEAOYULQ-UHFFFAOYNA-N
SMILESCC1CCC(C(=O)C1)C(C)(C)S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4920-1.4970 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only