Perfluorodecanoic acid, 97%
Perfluorodecanoic acid, 97%
Perfluorodecanoic acid, 97%
Thermo Scientific Chemicals

Perfluorodecanoic acid, 97%

CAS: 335-76-2 | C10HF19O2 | 514.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22292.06
Price (USD)
61.65
Online exclusive
68.20
Save 6.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Online exclusive
68.20
Save 6.55
Each
Chemical Identifiers
CAS335-76-2
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0% (GC and/or titration)
Appearance (Color)White to cream
FormPowder or crystalline powder or Flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only