Ethyl 2-ethylacetoacetate, 95%
Ethyl 2-ethylacetoacetate, 95%
Ethyl 2-ethylacetoacetate, 95%
Thermo Scientific Chemicals

Ethyl 2-ethylacetoacetate, 95%

CAS: 607-97-6 | C8H14O3 | 158.197 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22295.14
also known as B22295-14
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS607-97-6
IUPAC Nameethyl 2-ethyl-3-oxobutanoate
Molecular FormulaC8H14O3
InChI KeyOKANYBNORCUPKZ-UHFFFAOYNA-N
SMILESCCOC(=O)C(CC)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Refractive Index1.4180-1.4240 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only