2-Bromo-2',4'-dimethoxyacetophenone, 98%
2-Bromo-2',4'-dimethoxyacetophenone, 98%
2-Bromo-2',4'-dimethoxyacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-2',4'-dimethoxyacetophenone, 98%

CAS: 60965-26-6 | C10H11BrO3 | 259.099 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22300.06
also known as B22300-06
Price (USD)
255.65
Special offer
Online exclusive
Ends: 30-Jun-2026
300.00
Save 44.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
255.65
Special offer
Online exclusive
Ends: 30-Jun-2026
300.00
Save 44.35 (15%)
Each
Chemical Identifiers
CAS60965-26-6
IUPAC Name2-bromo-1-(2,4-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H11BrO3
InChI KeyPKVBZABQCCQHLD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(=O)CBr)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)102.0-108.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only