2-Hexanoylfuran, 99%
2-Hexanoylfuran, 99%
2-Hexanoylfuran, 99%
Thermo Scientific Chemicals

2-Hexanoylfuran, 99%

CAS: 14360-50-0 | C10H14O2 | 166.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22306.06
also known as B22306-06
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.80
Save 8.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Special offer
Online exclusive
Ends: 28-Dec-2026
53.80
Save 8.15
Each
Chemical Identifiers
CAS14360-50-0
IUPAC Name1-(furan-2-yl)hexan-1-one
Molecular FormulaC10H14O2
InChI KeyYUAYWSBSIJVIBS-UHFFFAOYSA-N
SMILESCCCCCC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4875-1.4905 @ 20?C
Assay (GC)≥98.5%
Appearance (Color)Clear pale yellow to yellow
FormLiquid
2-Hexanoylfuran is used as a food additives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexanoylfuran is used as a food additives

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. M. M. El-Dessouky.; I. Gar Al-Alm Rashed. Radiolysis of 2-propanol in presence of 2-hexanoylfuran. Journal of Radioanalytical and Nuclear Chemistry. 1988, 120, (2), 361-368.
  2. Frederick Peter Sargent.; Edward Michael Gardy. The Detection of Alkoxy and Other Radicals in the Gamma Radiolysis of Alcohols by an Electron Spin Resonance and Spin Trapping Method. Canadian Journal of Chemistry.1974, 52, (21), 3645-3650.