1-(p-Toluenesulfonyl)indole, 95%
1-(p-Toluenesulfonyl)indole, 95%
1-(p-Toluenesulfonyl)indole, 95%
Thermo Scientific Chemicals

1-(p-Toluenesulfonyl)indole, 95%

CAS: 31271-90-6 | C15H13NO2S | 271.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22320.06
also known as B22320-06
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS31271-90-6
IUPAC Name1-(4-methylbenzenesulfonyl)-1H-indole
Molecular FormulaC15H13NO2S
InChI KeyJNRRPYFLDADLJW-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)N1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)79.0-91.0?C
Assay (GC)≥94.0%
Appearance (Color)White to pale cream
FormPowder
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(p-Toluenesulfonyl)indole, is synthesized and used as a stable equivalent of 4-mercaptoindoles, also used in medicinal field for making drug and tablets.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Terry V. Hughes.; Michael P. Cava. Electrophilic Cyanations Using 1-Cyanobenzotriazole: sp2 and sp Carbanions. J. Org. Chem. 1999, 64, (1), 313-315.
  2. Gerald S. Ponticello.; John J. Baldwin. Useful synthesis of 4-substituted indoles. J. Org. Chem.1979, 44, (22), 4003-4005.