Pentafluorothiophenol, 97%
Pentafluorothiophenol, 97%
Pentafluorothiophenol, 97%
Thermo Scientific Chemicals

Pentafluorothiophenol, 97%

CAS: 771-62-0 | C6HF5S | 200.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22323.06
also known as B22323-06
Price (USD)
89.10
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
89.10
Each
Chemical Identifiers
CAS771-62-0
IUPAC Namepentafluorobenzene-1-thiol
Molecular FormulaC6HF5S
InChI KeyUVAMFBJPMUMURT-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(S)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4610-1.4660 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid
Pentafluorothiophenol is used as a fluorinated building block

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorothiophenol is used as a fluorinated building block

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. V. Srini J. De Mel.; Rajesh Kumar.; John P. Oliver. Synthesis and characterization of alkyl aluminum compounds of pentafluorothiophenol. Crystal and molecular structure of [Me2Al(.mu.-SC6F5)]2. Organometallics. 1990, 9, (4), 1303-1307.
  2. Brian S Hammes.; Carl J Carrano. The structure and characterization of zinc heteroscorpionate complexes containing pentafluorothiophenol. Inorganica Chimica Acta.2000, 300-302, 427-433.