trans-2-Aminocyclohexanol hydrochloride, 97+%
trans-2-Aminocyclohexanol hydrochloride, 97+%
trans-2-Aminocyclohexanol hydrochloride, 97+%
Thermo Scientific Chemicals

trans-2-Aminocyclohexanol hydrochloride, 97+%

CAS: 5456-63-3 | C6H13ClNO | 150.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22324.14
also known as B22324-14
Price (USD)
276.65
Online Exclusive
307.00
Save 30.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
276.65
Online Exclusive
307.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS5456-63-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or granules
Assay (Titration ex Chloride)≥97.0 to ≤103.0% (Dry wt. basis)
Water Content (Karl Fischer Titration)≤2.0%
Melting Point (clear melt)168.0-179.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only