Di-n-hexyl sulfide, 97%
Di-n-hexyl sulfide, 97%
Di-n-hexyl sulfide, 97%
Thermo Scientific Chemicals

Di-n-hexyl sulfide, 97%

CAS: 6294-31-1 | C12H26S | 202.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22325.14
also known as B22325-14
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.30
Save 8.65 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.30
Save 8.65 (15%)
Each
Chemical Identifiers
CAS6294-31-1
IUPAC Name1-(hexylsulfanyl)hexane
Molecular FormulaC12H26S
InChI KeyLHNRHYOMDUJLLM-UHFFFAOYSA-N
SMILESCCCCCCSCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4560-1.4610 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only