4-Phenoxybenzaldehyde, 98%
4-Phenoxybenzaldehyde, 98%
4-Phenoxybenzaldehyde, 98%
Thermo Scientific Chemicals

4-Phenoxybenzaldehyde, 98%

CAS: 67-36-7 | C13H10O2 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22329.03
also known as B22329-03
Price (USD)
34.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.40
Each
Chemical Identifiers
CAS67-36-7
IUPAC Name4-phenoxybenzaldehyde
Molecular FormulaC13H10O2
InChI KeyQWLHJVDRPZNVBS-UHFFFAOYSA-N
SMILESO=CC1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Free acid (titration)<1.5%
Refractive Index1.6090-1.6130 @ 20?C
Appearance (Color)Clear colorless to pale yellow
4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Reuben D. Rieke.; Seung-Hoi Kim. Antimicrobial activity of wool fabric treated with curcumin. Dyes and Pigments. 2005, 65, (2), 157-161.