1-Phenylsemicarbazide, 99%
1-Phenylsemicarbazide, 99%
1-Phenylsemicarbazide, 99%
Thermo Scientific Chemicals

1-Phenylsemicarbazide, 99%

CAS: 103-03-7 | C7H9N3O | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B22331.14
also known as B22331-14
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS103-03-7
IUPAC Name(phenylamino)urea
Molecular FormulaC7H9N3O
InChI KeyAVKHCKXGKPAGEI-UHFFFAOYSA-N
SMILESNC(=O)NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale orange
FormCrystals or powder or crystalline powder
Melting Point (clear melt)168.0-178.0?C
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only