9-Vinylanthracene, 97%
9-Vinylanthracene, 97%
9-Vinylanthracene, 97%
9-Vinylanthracene, 97%
Thermo Scientific Chemicals

9-Vinylanthracene, 97%

CAS: 2444-68-0 | C16H12 | 204.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22333.06
also known as B22333-06
Price (USD)
276.65
Online Exclusive
307.00
Save 30.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
276.65
Online Exclusive
307.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2444-68-0
IUPAC Name9-ethenylanthracene
Molecular FormulaC16H12
InChI KeyOGOYZCQQQFAGRI-UHFFFAOYSA-N
SMILESC=CC1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)59.0-68.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only