5-Methyl-2-hepten-4-one, predominantly trans, 99%
5-Methyl-2-hepten-4-one, predominantly trans, 99%
5-Methyl-2-hepten-4-one, predominantly trans, 99%
Thermo Scientific Chemicals

5-Methyl-2-hepten-4-one, predominantly trans, 99%

CAS: 81925-81-7 | C8H14O | 126.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number B22335.04
also known as B22335-04
Price (USD)
58.65
Online exclusive
65.30
Save 6.65
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
58.65
Online exclusive
65.30
Save 6.65
Each
Chemical Identifiers
CAS81925-81-7
IUPAC Name(2E)-5-methylhept-2-en-4-one
Molecular FormulaC8H14O
InChI KeyARJWAURHQDJJAC-GQCTYLIANA-N
SMILESCCC(C)C(=O)C=CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4420-1.4450 @ 20°C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only