4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Thermo Scientific Chemicals

4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

CAS: 637-69-4 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B22353.06
also known as B22353-06
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.30
Save 11.65 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.30
Save 11.65 (15%)
Each
Chemical Identifiers
CAS637-69-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.5585-1.5635 @ 20°C
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

Solubility
Slightly miscible with methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Perkowski, A. J.; You, W.; Nicewicz, D. A. Visible Light Photoinitiated Metal-Free Living Cationic Polymerization of 4-Methoxystyrene. J. Am. Chem. Soc. 2015, 137 (24), 7580-7583.
  2. Miyazawa, K.; Koike, T.; Akita, M. Regiospecific Intermolecular Aminohydroxylation of Olefins by Photoredox Catalysis. Chem. Eur. J. 2015, 21 (33), 11677-11680.